Gene Transcripts

Gene Symbol
Cdh9
Entrez Gene ID
29163
Gene Name
cadherin 9, type 2, T1-cadherin
Gene Aliases
Chromosome Location
Chr. 2: 66938794 - 67078478 on Build Rnor_6.0
UniGene ID
Rn.222436
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168630.1 NP_001162101.1 10 1721
XM_006232076.2 XP_006232138.1 12 1945

View Full Product Details