Gene Transcripts
- Gene Symbol
- Btg1
- Entrez Gene ID
- 29618
- Gene Name
- B-cell translocation gene 1, anti-proliferative
- Gene Aliases
- Chromosome Location
- Chr. 7: 37812831 - 37815088 on Build Rnor_6.0
- UniGene ID
- Rn.1000
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017258.1 |
NP_058954.1
|
2 |
418 |
|
GenBank mRNA |
L26268.1 |
- |
2 |
418 |
|
View Full Product Details