Gene Transcripts
- Gene Symbol
- SNRNP70
- Entrez Gene ID
- 6625
- Gene Name
- small nuclear ribonucleoprotein U1 subunit 70
- Gene Aliases
- RNPU1Z, RPU1, SNRP70, Snp1, U1-70K, U170K, U1AP, U1RNP
- Chromosome Location
- Chr. 19: 49085140 - 49108613 on Build GRCh38
- UniGene ID
- Hs.467097
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301069.1 |
NP_001287998.1
|
6, 7 |
893 |
|
NM_003089.5 |
NP_003080.2
|
6, 7 |
893 |
|
GenBank mRNA |
AK096783.1 |
- |
6, 7 |
582 |
|
AK293120.1 |
- |
6, 7 |
612 |
|
AL560281.3 |
- |
6, 7 |
581 |
|
BC001315.1 |
- |
6, 7 |
566 |
3454479 |
M22636.1 |
- |
6, 7 |
557 |
|
X06812.1 |
- |
4, 5 |
241 |
|
X06815.1 |
- |
6, 7 |
537 |
|
X84841.1 |
- |
5, 6 |
386 |
|
View Full Product Details