Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031149.1 | NP_112411.1 | 6 | 456 | |
GenBank mRNA | AB000491.1 | - | 6 | 456 | |
BC058462.1 | - | 6 | 452 | 6891005 | |
D83521.1 | - | 6 | 474 | ||
FQ213263.1 | - | 6 | 473 | ||
FQ233831.1 | - | 6 | 459 |