Gene Transcripts
- Gene Symbol
- Itgae
- Entrez Gene ID
- 83577
- Gene Name
- integrin, alpha E
- Gene Aliases
- Chromosome Location
- Chr. 10: 59652549 - 59711793 on Build Rnor_6.0
- UniGene ID
- Rn.29975
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031768.2 |
NP_113956.2
|
21 |
2648 |
|
GenBank mRNA |
AF020046.1 |
- |
21 |
2617 |
|
View Full Product Details