Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001287742.1 | NP_001274671.1 | 10 | 2595 | |
NM_001287743.1 | NP_001274672.1 | 9 | 2397 | ||
NM_001287744.1 | NP_001274673.1 | 8 | 2224 | ||
NM_001287745.1 | NP_001274674.1 | 9 | 2303 | ||
NM_001287747.1 | NP_001274676.1 | 8 | 2260 | ||
NM_001287748.1 | NP_001274677.1 | 8 | 1922 | ||
NM_001287749.1 | NP_001274678.1 | 8 | 2160 | ||
NM_001287750.1 | NP_001274679.1 | 7 | 2162 | ||
NM_001287751.1 | NP_001274680.1 | 7 | 2034 | ||
NM_001287756.1 | NP_001274685.1 | 5 | 1619 | ||
NM_004462.4 | NP_004453.3 | 8 | 2106 | ||
XM_017013217.1 | XP_016868706.1 | 10 | 2887 | ||
GenBank mRNA | X69141.1 | - | 8 | 1975 |