Gene Transcripts
- Gene Symbol
- Csn1s2b
- Entrez Gene ID
- 286759
- Gene Name
- casein alpha s2-like B
- Gene Aliases
- Csnd
- Chromosome Location
- Chr. 14: 21899954 - 21916068 on Build Rnor_6.0
- UniGene ID
- Rn.91144
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173106.1 |
NP_775129.1
|
2, 3 |
115 |
|
GenBank mRNA |
AY154894.1 |
- |
2, 3 |
115 |
|
View Full Product Details