Gene Transcripts

Gene Symbol
Vom2r-ps45
Entrez Gene ID
286982
Gene Name
vomeronasal 2 receptor, pseudogene 45
Gene Aliases
Chromosome Location
Chr. 1: 69496423 - 69522162 on Build Rnor_6.0
UniGene ID
Rn.10811
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF016183.1 - 453

View Full Product Details