Gene Transcripts

Gene Symbol
Ptrh2
Entrez Gene ID
287593
Gene Name
peptidyl-tRNA hydrolase 2
Gene Aliases
RGD1306819
Chromosome Location
Chr. 10: 74002151 - 74012182 on Build Rnor_6.0
UniGene ID
Rn.13456
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013860.1 NP_001013882.1 2 415
GenBank mRNA BC083801.1 - 2 415

View Full Product Details