Gene Transcripts

Gene Symbol
Galk1
Entrez Gene ID
287835
Gene Name
galactokinase 1
Gene Aliases
Galk, Glk
Chromosome Location
Chr. 10: 104560322 - 104564499 on Build Rnor_6.0
UniGene ID
Rn.198917
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008282.1 NP_001008283.1 4 622
GenBank mRNA BC085919.1 - 4 622
FQ219172.1 - 4 636
FQ234739.1 - 4 595

View Full Product Details