Gene Transcripts

Gene Symbol
Evc
Entrez Gene ID
289712
Gene Name
Ellis van Creveld syndrome
Gene Aliases
Chromosome Location
Chr. 14: 78213601 - 78253266 on Build Rnor_6.0
UniGene ID
Rn.25111
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170439.1 NP_001163910.1 15 2330
XM_006251090.2 XP_006251152.1 15 2364
XM_006251091.1 XP_006251153.1 12 1827

View Full Product Details