Gene Transcripts

Gene Symbol
TULP3
Entrez Gene ID
7289
Gene Name
tubby like protein 3
Gene Aliases
TUBL3
Chromosome Location
Chr. 12: 2890867 - 2941140 on Build GRCh38
UniGene ID
Hs.655333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160408.1 NP_001153880.1 2, 3 170
NM_003324.4 NP_003315.2 2, 3 170
GenBank mRNA AF045583.1 - 2, 3 134
AK024246.1 - 2, 3 170
BC032587.1 - 2, 3 143 5501868

View Full Product Details