Gene Transcripts

Gene Symbol
Col4a1
Entrez Gene ID
290905
Gene Name
collagen, type IV, alpha 1
Gene Aliases
Chromosome Location
Chr. 16: 83522162 - 83632153 on Build Rnor_6.0
UniGene ID
Rn.53801
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135009.1 NP_001128481.1 48 4559
GenBank mRNA BC089096.1 - 2 140

View Full Product Details