Gene Transcripts

Gene Symbol
Gadd45g
Entrez Gene ID
291005
Gene Name
growth arrest and DNA-damage-inducible, gamma
Gene Aliases
Chromosome Location
Chr. 17: 13391474 - 13393219 on Build Rnor_6.0
UniGene ID
Rn.16950
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077640.1 NP_001071108.1 3 255
GenBank mRNA AB020978.1 - 3 255
BC168643.1 - 3 255 9092341
FQ218856.1 - 3 273

View Full Product Details