Gene Transcripts

Gene Symbol
Mybpc2
Entrez Gene ID
292879
Gene Name
myosin binding protein C, fast-type
Gene Aliases
Chromosome Location
Chr. 1: 100506704 - 100530190 on Build Rnor_6.0
UniGene ID
Rn.137668
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106257.1 NP_001099727.1 19 2198
XM_006229002.2 XP_006229064.1 19 2160

View Full Product Details