Gene Transcripts

Gene Symbol
Adamts17
Entrez Gene ID
293004
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif, 17
Gene Aliases
Chromosome Location
Chr. 1: 127802756 - 128126764 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229360.2 XP_006229422.1 10 1674
XM_218753.8 XP_218753.6 10 1673

View Full Product Details