Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001303543.1 | NP_001290472.1 | 6 | 752 | |
NM_001303544.1 | NP_001290473.1 | 7 | 690 | ||
NM_001303545.1 | NP_001290474.1 | 6 | 597 | ||
NM_003372.6 | NP_003363.1 | 6 | 601 | ||
GenBank mRNA | AK301646.1 | - | 7 | 690 | |
AK313420.1 | - | 6 | 569 | ||
BC046094.1 | - | 6 | 554 | 6058145 | |
BT019604.1 | - | 6 | 542 | ||
CR541932.1 | - | 5 | 431 | ||
U56833.1 | - | 5 | 450 | ||
U96759.1 | - | 6 | 749 | ||
Y17394.1 | - | 6 | 554 |