Gene Transcripts

Gene Symbol
Hmx2
Entrez Gene ID
293538
Gene Name
H6 family homeobox 2
Gene Aliases
RGD1565366
Chromosome Location
Chr. 1: 203515605 - 203523551 on Build Rnor_6.0
UniGene ID
Rn.102367
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106303.1 NP_001099773.1 3 867
XM_008759880.1 XP_008758102.1 3 1448

View Full Product Details