Gene Transcripts

Gene Symbol
Dtx4
Entrez Gene ID
293774
Gene Name
deltex 4, E3 ubiquitin ligase
Gene Aliases
LRRGT00037
Chromosome Location
Chr. 1: 228730081 - 228759781 on Build Rnor_6.0
UniGene ID
Rn.23314
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047855.1 NP_001041320.1 6 610
XM_006231114.2 XP_006231176.1 7 2431
GenBank mRNA AY383692.1 - 6 610

View Full Product Details