Gene Transcripts

Gene Symbol
Supv3l1
Entrez Gene ID
294385
Gene Name
suppressor of var1, 3-like 1 (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 20: 32057530 - 32080170 on Build Rnor_6.0
UniGene ID
Rn.19830
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012462.1 NP_001012480.1 15 1952
GenBank mRNA BC089883.1 - 15 1952

View Full Product Details