Gene Transcripts

Gene Symbol
Tgm6
Entrez Gene ID
296152
Gene Name
transglutaminase 6
Gene Aliases
RGD1565348
Chromosome Location
Chr. 3: 122644183 - 122680049 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762236.1 XP_008760458.1 2 1699

View Full Product Details