Gene Transcripts

Gene Symbol
Tmem8c
Entrez Gene ID
296601
Gene Name
transmembrane protein 8C
Gene Aliases
RGD1306074
Chromosome Location
Chr. 3: 5608243 - 5617689 on Build Rnor_6.0
UniGene ID
Rn.162652
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134517.1 NP_001127989.1 1 305
XM_008761635.1 XP_008759857.1 1 307

View Full Product Details