Gene Transcripts

Gene Symbol
Epha10
Entrez Gene ID
298528
Gene Name
EPH receptor A10
Gene Aliases
RGD1562679
Chromosome Location
Chr. 5: 142875683 - 142910537 on Build Rnor_6.0
UniGene ID
Rn.118682
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135709.1 NP_001129181.1 16 2645
XM_006238858.2 XP_006238920.1 14 2637

View Full Product Details