Gene Transcripts

Gene Symbol
Ift27
Entrez Gene ID
300062
Gene Name
intraflagellar transport 27
Gene Aliases
Rabl4
Chromosome Location
Chr. 7: 119393384 - 119409722 on Build Rnor_6.0
UniGene ID
Rn.9036
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130495.1 NP_001123967.1 2 393
XM_006241931.2 XP_006241993.1 2 405
GenBank mRNA BC168150.1 - 403

View Full Product Details