Gene Transcripts

Gene Symbol
Lysmd2
Entrez Gene ID
300839
Gene Name
LysM, putative peptidoglycan-binding, domain containing 2
Gene Aliases
RGD1304585
Chromosome Location
Chr. 8: 82559566 - 82577029 on Build Rnor_6.0
UniGene ID
Rn.7432
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106839.1 NP_001100309.1 1, 2 371
XM_006243405.2 XP_006243467.1 1, 2 1264
GenBank mRNA BC167017.1 - 1, 2 263

View Full Product Details