Gene Transcripts

Gene Symbol
Igsf1
Entrez Gene ID
302822
Gene Name
immunoglobulin superfamily, member 1
Gene Aliases
Inhbp
Chromosome Location
Chr. X: 136792637 - 136808107 on Build Rnor_6.0
UniGene ID
Rn.23738
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175763.2 NP_786939.1 8 1584
XM_006257557.2 XP_006257619.1 9 1705
XM_006257558.1 XP_006257620.1 8 1655
XM_006257559.1 XP_006257621.1 8 1658
XM_006257560.1 XP_006257622.1 8 1658
XM_006257561.1 XP_006257623.1 8 1658
XM_008773563.1 XP_008771785.1 8 1658
XM_008773564.1 XP_008771786.1 1 661
GenBank mRNA AF322216.1 - 8 1584

View Full Product Details