Gene Transcripts

Gene Symbol
PSCA
Entrez Gene ID
8000
Gene Name
prostate stem cell antigen
Gene Aliases
PRO232
Chromosome Location
Chr. 8: 142670308 - 142682727 on Build GRCh38
UniGene ID
Hs.652235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005672.4 NP_005663.2 3 843
NR_033343.1 3 996
GenBank mRNA BC048808.1 - 3 793

View Full Product Details