Gene Transcripts

Gene Symbol
Rc3h1
Entrez Gene ID
680586
Gene Name
ring finger and CCCH-type domains 1
Gene Aliases
RGD1563512
Chromosome Location
Chr. 13: 78723105 - 78794663 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769698.1 XP_008767920.1 3 634

View Full Product Details