Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008764496.1 | XP_008762718.1 | 6 | 1338 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001191930.1 | NP_001178859.1 | 3 | 624 | |
XM_006250222.1 | XP_006250284.1 | 3 | 619 | ||
XM_006250223.1 | XP_006250285.1 | 3 | 619 |