Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006011.3 | NP_006002.1 | 2 | 303 | |
XM_017022642.1 | XP_016878131.1 | 2 | 466 | ||
GenBank mRNA | AK313924.1 | - | 2 | 251 | |
AW163487.1 | - | 303 | |||
BC069584.1 | - | 2 | 172 | ||
BC096202.3 | - | 2 | 172 | 40004644 | |
BC096203.3 | - | 2 | 172 | 40004645 | |
BC096204.3 | - | 2 | 172 | 40004646 | |
U33551.1 | - | 2 | 148 | ||
U82762.1 | - | 2 | 173 |