Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006252404.2 | XP_006252466.1 | 5 | 832 | |
XM_006252405.2 | XP_006252467.1 | 5 | 785 | ||
XM_006252406.2 | XP_006252468.1 | 5 | 617 | ||
XM_006252407.2 | XP_006252469.1 | 5 | 832 | ||
XM_224451.8 | XP_224451.3 | 5 | 658 | ||
GenBank mRNA | FQ210517.1 | - | 820 |