Gene Transcripts

Gene Symbol
Nhlrc2
Entrez Gene ID
307986
Gene Name
NHL repeat containing 2
Gene Aliases
Chromosome Location
Chr. 1: 277355619 - 277415940 on Build Rnor_6.0
UniGene ID
Rn.161909
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107444.1 NP_001100914.1 5, 6 1469
XM_006231645.2 XP_006231707.1 5, 6 1469
XM_008760536.1 XP_008758758.1 5, 6 1243

View Full Product Details