Gene Transcripts

Gene Symbol
Vom2r6
Entrez Gene ID
308257
Gene Name
vomeronasal 2 receptor, 6
Gene Aliases
RGD1562697
Chromosome Location
Chr. 1: 69052895 - 69064777 on Build Rnor_6.0
UniGene ID
Rn.217639
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099459.1 NP_001092929.1 380

View Full Product Details