Gene Transcripts

Gene Symbol
Orai3
Entrez Gene ID
309000
Gene Name
ORAI calcium release-activated calcium modulator 3
Gene Aliases
RGD1306538
Chromosome Location
Chr. 1: 199217504 - 199222363 on Build Rnor_6.0
UniGene ID
Rn.42186
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014024.1 NP_001014046.1 2 933
GenBank mRNA BC079355.1 - 2 933
EU644751.1 - 2 780

View Full Product Details