Gene Transcripts

Gene Symbol
Mrpl43
Entrez Gene ID
309440
Gene Name
mitochondrial ribosomal protein L43
Gene Aliases
Chromosome Location
Chr. 1: 264755537 - 264756404 on Build Rnor_6.0
UniGene ID
Rn.225372
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107598.1 NP_001101068.1 2 209
XM_008760428.1 XP_008758650.1 2 242

View Full Product Details