Gene Transcripts

Gene Symbol
Shoc2
Entrez Gene ID
309548
Gene Name
soc-2 suppressor of clear homolog (C. elegans)
Gene Aliases
Chromosome Location
Chr. 1: 274700621 - 274751195 on Build Rnor_6.0
UniGene ID
Rn.156055
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013155.1 NP_001013173.1 835
GenBank mRNA BC079032.1 - 835

View Full Product Details