Gene Transcripts

Gene Symbol
Mast4
Entrez Gene ID
100912235
Gene Name
microtubule associated serine/threonine kinase family member 4
Gene Aliases
RGD1310139
Chromosome Location
Chr. 2: 32442285 - 32519052 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760713.1 XP_008758935.1 1481

Gene Symbol
NEWGENE_1310139
Entrez Gene ID
103690045
Gene Name
microtubule associated serine/threonine kinase family member 4
Gene Aliases
Mast4
Chromosome Location
Chr. 2: 32857576 - 33026430 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760704.1 XP_008758926.1 13 2829
XM_008760705.1 XP_008758927.1 13 2829

View Full Product Details