Gene Transcripts

Gene Symbol
Ext2
Entrez Gene ID
311215
Gene Name
exostosin glycosyltransferase 2
Gene Aliases
Chromosome Location
Chr. 3: 82602785 - 82734562 on Build Rnor_6.0
UniGene ID
Rn.2878
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107751.1 NP_001101221.1 5, 6 1153
XM_006234576.2 XP_006234638.2 5, 6 1182
GenBank mRNA BC166748.1 - 5, 6 1140

View Full Product Details