Gene Transcripts

Gene Symbol
Nat10
Entrez Gene ID
311257
Gene Name
N-acetyltransferase 10 (GCN5-related)
Gene Aliases
RGD1306717
Chromosome Location
Chr. 3: 93658119 - 93698136 on Build Rnor_6.0
UniGene ID
Rn.40316
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107756.1 NP_001101226.1 5 633
GenBank mRNA BC169067.1 - 6 741

View Full Product Details