Gene Transcripts

Gene Symbol
Tomm34
Entrez Gene ID
311621
Gene Name
translocase of outer mitochondrial membrane 34
Gene Aliases
Chromosome Location
Chr. 3: 160448294 - 160465143 on Build Rnor_6.0
UniGene ID
Rn.16749
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044244.2 NP_001037709.1 5 627
XM_006235585.2 XP_006235647.1 5 902
GenBank mRNA BC105768.1 - 5 628

View Full Product Details