Gene Transcripts

Gene Symbol
Plxnd1
Entrez Gene ID
312652
Gene Name
plexin D1
Gene Aliases
Chromosome Location
Chr. 4: 147854309 - 147894173 on Build Rnor_6.0
UniGene ID
Rn.21111
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107881.1 NP_001101351.1 17 3109
XM_006237035.2 XP_006237097.1 17 3377

View Full Product Details