Gene Transcripts

Gene Symbol
Gprc5a
Entrez Gene ID
312790
Gene Name
G protein-coupled receptor, class C, group 5, member A
Gene Aliases
Rai3
Chromosome Location
Chr. 4: 168832937 - 168851650 on Build Rnor_6.0
UniGene ID
Rn.23566
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079890.1 NP_001073359.1 4 1126
GenBank mRNA BC086372.1 - 3 501
BC128767.1 - 4 1126 7124284

View Full Product Details