Gene Transcripts

Gene Symbol
Etnk1
Entrez Gene ID
312828
Gene Name
ethanolamine kinase 1
Gene Aliases
Chromosome Location
Chr. 4: 177402904 - 177447047 on Build Rnor_6.0
UniGene ID
Rn.7630
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107894.1 NP_001101364.1 3 772
XM_006237623.1 XP_006237685.1 3 767

View Full Product Details