Gene Transcripts

Gene Symbol
Stk11
Entrez Gene ID
314621
Gene Name
serine/threonine kinase 11
Gene Aliases
Lkb1
Chromosome Location
Chr. 7: 12440751 - 12457513 on Build Rnor_6.0
UniGene ID
Rn.12052
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108069.1 NP_001101539.1 2, 3 1201
XM_006240910.1 XP_006240972.1 2, 3 1196
XM_008765096.1 XP_008763318.1 2, 3 1196

View Full Product Details