Gene Transcripts

Gene Symbol
Cdk17
Entrez Gene ID
314743
Gene Name
cyclin-dependent kinase 17
Gene Aliases
Pctaire2, Pctk2
Chromosome Location
Chr. 7: 34001506 - 34079616 on Build Rnor_6.0
UniGene ID
Rn.17706
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108082.2 NP_001101552.2 12 1567
XM_006241253.2 XP_006241315.1 12 1817
GenBank mRNA AB005540.1 - 12 1532

View Full Product Details