Gene Transcripts

Gene Symbol
Hoxc10
Entrez Gene ID
315338
Gene Name
homeo box C10
Gene Aliases
Chromosome Location
Chr. 7: 144577512 - 144582802 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750414.3 XP_003750462.1 2 971
GenBank mRNA S71286.1 - 1 44

View Full Product Details