Gene Transcripts
- Gene Symbol
- Sik2
- Entrez Gene ID
- 315649
- Gene Name
- salt-inducible kinase 2
- Gene Aliases
- RGD1559698, Snf1lk2
- Chromosome Location
- Chr. 8: 55311700 - 55408607 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DQ188032.1 |
- |
|
389 |
|
View Full Product Details