Gene Transcripts
- Gene Symbol
- Hecw2
- Entrez Gene ID
- 316395
- Gene Name
- HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2
- Gene Aliases
- Chromosome Location
- Chr. 9: 60451476 - 60672246 on Build Rnor_6.0
- UniGene ID
- Rn.198842
- Species
- Rattus norvegicus
View Full Product Details