Gene Transcripts

Gene Symbol
Myom1
Entrez Gene ID
316740
Gene Name
myomesin 1
Gene Aliases
Chromosome Location
Chr. 9: 119353840 - 119469196 on Build Rnor_6.0
UniGene ID
Rn.15504
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191584.1 NP_001178513.1 5 1001

View Full Product Details