Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320099.1 | NP_001307028.1 | 2 | 437 | |
NM_001320100.1 | NP_001307029.1 | 2 | 227 | ||
NM_001320101.1 | NP_001307030.1 | 2 | 227 | ||
NM_003805.4 | NP_003796.1 | 2 | 227 | ||
NR_135147.1 | 2 | 227 | |||
XM_017020142.1 | XP_016875631.1 | 2 | 256 | ||
XM_017020143.1 | XP_016875632.1 | 2 | 250 | ||
XM_017020144.1 | XP_016875633.1 | 2 | 250 | ||
XM_017020145.1 | XP_016875634.1 | 2 | 249 | ||
GenBank mRNA | AB451280.1 | - | 1 | 119 | |
AB451412.1 | - | 1 | 119 | ||
AK294986.1 | - | 2 | 215 | ||
AK296225.1 | - | 2 | 225 | ||
AK314861.1 | - | 2 | 207 | ||
BC017042.1 | - | 2 | 199 | 3849848 | |
BC037905.1 | - | 2 | 441 | 5276510 | |
BT009837.1 | - | 1 | 119 | ||
CR407643.1 | - | 1 | 119 | ||
HY113325.1 | - | 2 | 228 | ||
U79115.1 | - | 2 | 219 | ||
U84388.1 | - | 2 | 156 |